Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MRP1 Rat Monoclonal Antibody

Supplier: Enzo Life Sciences
801-007-C250 801-007-C125
89153-172EA 635 USD InStock
89153-172 89153-174
Anti-MRP1 Rat Monoclonal Antibody
Antibodies
Host: Rat, Isotype: IgG2a

  • For Immunocytochemistry, Immunohistochemistry, Western Blot

Recognizes the internal epitope (G238SDLWSLNKE247) of human and mouse MRP1, overexpressed in various human non-P-glycoprotein MDR tumor cell lines. Does not cross-react with human P-glycoprotein, MDR3 P-glycoprotein or human MRP2 (cMOAT) to MRP 5.

Type: Primary
Antigen: MRP1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty