Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MRP2 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
801-037-C250 801-037-C125
89153-252EA 654 USD InStock
89153-252 89153-254
Anti-MRP2 Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG2b

  • For Immunocytochemistry, Immunohistochemistry, Western Blot

Recognizes an internal epitope of human, mouse and rat MRP2. Does not cross-react with human P-glycoprotein, MDR3 P-glycoprotein, MRP1, MRP3 or MRP5. Does not recognize mouse MRP2 in immunohistochemistry.

Type: Primary
Antigen: MRP2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty