Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GCDFP15 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
801-051-L001
89153-268EA 1250 USD InStock
89153-268
Anti-GCDFP15 Mouse Monoclonal Antibody
Antibodies
Gross cystic disease is a common premenopausal disorder in which gross cysts are the predominant pathologic lesion. It is characterized by production of a fluid secretion which accumulates in the breast cysts. Gross cystic disease fluid is a pathologic secretion from breast composed of several glycoproteins, including a unique 15kDa monomer protein, GCDFP-15. The cells within the body that produce GCDFP-15 appear to be restricted primarily to those with apocrine function. Studies have found GCDFP-15 to be a highly specific and sensitive marker for breast cancer.
GCDFP-15 is expressed by apocrine sweat glands, eccrine glands (variable), minor salivary glands, bronchial glands, metaplasmic epithelium of the breast, benign sweat gland tumors of the skin and the serous cells of the submandibular gland. Breast carcinomas (primary and metastatic lesions) with apocrine features express GCDFP-15. GCDFP-15 is positive in extramammary Paget's Disease. Other tumors tested are negative.

  • For Immunohistochemistry

Recognizes human GCDFP-15.

Type: Primary
Antigen: GCDFP-15
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty