Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD44 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
801-082-C100
89153-304EA 309 USD InStock
89153-304
Anti-CD44 Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For Western Blot

Recognizes an epitope encoded by exon v7 on the variant portion of human CD44.

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: 0
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty