Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-alpha Gal Epitope Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
ALX-801-090-1
89153-336EA 590 USD InStock
89153-336
Anti-alpha Gal Epitope Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgM

  • For ELISA, Flow Cytometry, Immunohistochemistry, Western Blot

Recognizes synthetic and naturally produced mouse, rat and pig Galalpha1-3Gal epitopes on glycoproteins and glycolipids. Does not cross-react with beta-Gal glycoproteins or BSA.

Type: Primary
Antigen: [delete]
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty