Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Nicotinic Acetylcholine Receptor Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
802-002-R100
89153-342EA 647 USD InStock
89153-342
Anti-Nicotinic Acetylcholine Receptor Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes the intracellular domain of skeletal muscle nAChR gamma and delta subunits in Torpedo californica and the delta subunit in mouse, rat, chicken, frog and fish. Detects bands of ~60kDa (gamma subunit ) and ~65kDa (delta subunit ) by Western blot. Under non-reducing conditions detects a band of ~130kDa (dimer of delta subunit). Does not cross-react with the alpha1 and beta1 subunits in muscle or the neuronal nAChRs.

Type: Primary
Antigen: NICOTINIC ACETYLCHOLINE RECEPTOR
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty