Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ProlActin Receptor Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
803-003-C100
89153-372EA 573 USD InStock
89153-372
Anti-ProlActin Receptor Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG2a

  • For Immunocytochemistry, Immunohistochemistry, Western Blot

Recognizes the ligand binding site of rat prolactin receptor. Detects a band of ~42kDa by Western blot. Does not cross-react with prolactin receptors from other species or with growth hormone receptor.

Reconstitute with 100µl PBS.

Type: Primary
Antigen: PROLACTIN RECEPTOR
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty