Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ESR1 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
803-004-C050
89153-374EA 811 USD InStock
89153-374
Anti-ESR1 Mouse Monoclonal Antibody
Antibodies
Steroid receptors are a large superfamily of ligand-dependent intracellular proteins that stimulate transcription of specific genes by binding to specific DNA sequences following activation by the appropriate hormone. Estrogen receptor (ER) has been classified into two distinct isoforms, α and β. ERα is known to interact in a ligand dependent and independent manner with several known coactivators including Receptor-Interacting Protein 140 (RIP140/ERAP140), ERAP160, and Steroid Receptor Coactivator-1 (SRC-1) to enhance transcriptional activity of target genes.

  • For Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes both the steroid occupied and unoccupied forms of human, mouse, rat, bovine and monkey ERalpha. Detects a band of ~67kDa by Western blot. Does not cross-react with androgen receptor, PR (progesterone receptor), GR (glucocorticoid receptor) or ERbeta. Does not inhibit binding of ER to nuclei and DNA.

Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty