Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ITGAV Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
803-304-C100
89153-448EA 406 USD InStock
89153-448
Anti-ITGAV Mouse Monoclonal Antibody
Antibodies
CD51 (integrin αV subunit) associates with CD61 (integrin β3 subunit) binding to the RGD sequence in extracellular matrix proteins like vitronectin, fibrinogen, thrombospondin, von Willebrand factor and osteopontin. The αV/β3 heterodimer is expressed in endothelial cells, leukocytes, NK cells, macrophages, neutrophils, platelets and several malignant tumor cells and seems to play a role in embryo implantation, angiogenesis, wound healing and cancer. αV/β3  is also highly expressed in osteoclasts being important for bone resorption.

  • For ELISA, Flow Cytometry, Immunoprecipitation

Recognizes human CD51 (alphaV integrin). Recognizes heterodimers alphaVbeta3, alphaVbeta1, alphaVbeta5 and alphaVbeta6. Does not cross-react with other species.

Type: Primary
Antigen: ITGAV
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty