Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DMPO Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
803-340-C100
89153-514EA 513 USD InStock
89153-514
Anti-DMPO Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG

  • For ELISA, Flow Cytometry, Immunoprecipitation, Western Blot

Recognizes DMPO (Prod. No. ALX-430-090), DMPO-octanoic acid, DMPO-protein adducts and DMPO-DNA adducts. Does not cross-react with non-adducted proteins or DNA. Does not cross-react with ovalbumin.

Type: Primary
Antigen: DMPO
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: pan

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty