Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PMCA Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
804-049-R100
89153-580EA 777 USD InStock
89153-580
Anti-PMCA Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG2a

  • For Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes all known (currently four) isoforms of human, rat, mouse, sheep, dog, monkey, rabbit, cat, chicken and amphibian plasma membrane PMCA ATPase. Detects a band of ~140kDa by Western blot. Also detects bands at ~95kDa and ~180kDa which may represent aggregation and/or natural proteolytic products of PMCA ATPase. Recognizes an epitope between aa 724-783 of human PMCA ATPase.

Type: Primary
Antigen: PMCA
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty