Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSPA8 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
804-067-R050
89153-608EA 797 USD InStock
89153-608
Anti-HSPA8 Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgM

  • For Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse, monkey and cat HSC70 (heat shock cognate protein 70). Detects a band of ~70kDa by Western blot.

Type: Primary
Antigen: HSPA8
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty