Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Clusterin Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
804-126-C100
89153-704EA 406 USD InStock
89153-704
Anti-Clusterin Mouse Monoclonal Antibody
Antibodies
Clusterin (SGP-2/sulfated glycoprotein-2; CLI; Apolipoprotein J (Apo J); TRPM-2) is the first identified extracellular mammalian chaperone and appears to be implicated in such diverse processes as sperm maturation, regulation of complement activation, programmed cell death, tissue remodelling and lipid transport. Increases in clusterin mRNA and protein have been consistently detected in apoptotic cell death paradigms, establishing clusterin gene expression as a prominent marker of apoptotic cell loss. A dual role has been shown for clusterin in that cells can be protected from damage and dead cells can be more easily removed after some types of cellular damage.

  • For Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human clusterin.

Type: Primary
Antigen: Clusterin
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty