Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BAFF Rat Monoclonal Antibody

Supplier: Enzo Life Sciences
804-131-C100
89153-716EA 483 USD InStock
89153-716
Anti-BAFF Rat Monoclonal Antibody
Antibodies
Host: Rat, Isotype: IgM

  • For Immunohistochemistry, Western Blot

Recognizes membrane-bound and soluble human and mouse (weak) BAFF.

Type: Primary
Antigen: BAFF
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgM
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty