Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG1 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
804-133-C100
89153-722EA 332 USD InStock
89153-722
Anti-IgG1 Mouse Monoclonal Antibody
Antibodies
For ELISA, Flow Cytometry, Immunoprecipitation, Western Blot

  • Endotoxin content: <0.1EU/µg antibody (LAL test; BioWhittaker)

For ELISA, Flow Cytometry, Immunoprecipitation, Western Blot

Reconstitute with 100µl sterile water.

Type: Control
Antigen: IgG1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Human
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty