Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DR6 Rat Monoclonal Antibody

Supplier: Enzo Life Sciences
804-150-C100
89153-754EA 481 USD InStock
89153-754
Anti-DR6 Rat Monoclonal Antibody
Antibodies
Apoptosis is induced by certain cytokines including TNF and Fas ligand (CD95L) of the TNF family through their death domain containing receptors, TNF-R1 and Fas (CD95). Several death receptors including DR3, TRAIL-R1, and TRAIL-R2 were identified. A death domain containing receptor in the TNF-R family was cloned and termed DR6 for death receptor-6. Like TNF-R1, DR6 interacts with death domain containing adapter molecule TRADD. Overexpression of DR6 induces apoptosis and activates NF-κB and JNK. DR6 is widely expressed in human tissues and cell lines. The ligand for DR6 has not been identified yet. T cells from DR6-deficient mice showed preference toward Th2 differentiation in vitro, suggesting that DR6, working through JNK functions to attenuate the Th2 response. Other studies of DR6-targeted mutant mice demonstrated that DR6 functions as a regulatory receptor for mediating CD4(+) T cell activation and maintaining proper immune responses.

Recognizes human DR6.

Type: Primary
Antigen: DR6
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty