Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-S Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
804-163-C100
89153-786EA 547 USD InStock
89153-786
Anti-S Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgM

  • For ELISA, Flow Cytometry, Immunoprecipitation, Western Blot

Recognizes a broad range of serine-phosphorylated proteins in crude cell extracts, interacting with phosphoserine surrounded by positive or neutral aa (e.g. substrates of PKA, PKB, PKC, PKG etc.).Please note that phosphoserine detection by MAbs always depends on the surrounding aa sequence.

Reconstitute with 1ml water (15 minutes at room temperature).

Ordering information: Positive control included.

Type: Primary
Antigen: S
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: pan

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty