Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Laminin-2 Rat Monoclonal Antibody

Supplier: Enzo Life Sciences
804-190-C100
89153-824EA 747 USD InStock
89153-824
Anti-Laminin-2 Rat Monoclonal Antibody
Antibodies
The laminins are a family of glycoproteins that provide an integral part of the structural scaffolding of basement membranes in almost every tissue. Each laminin is a heterotrimer assembled from α, β and γ chain subunits, secreted and incorporated into cell-associated extracellular matrices. The laminins can self-assemble, bind to other matrix macromolecules, and have unique and shared cell interactions mediated by integrins, dystroglycans, and other receptors. Through these interactions, laminins critically contribute to cell differentiation, cell shape and movement, maintenance of tissue phenotypes and promotion of tissue survival.

  • For ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation

Recognizes human and mouse laminin alpha2 (300kDa). Recognizes an N-terminal portion of the alpha2 chain that is deleted in congenital muscular dystrophies.

Reconstitute with 1ml distilled water (15 min, RT).

Type: Primary
Antigen: LAMININ-2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty