Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DNA Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
804-192-L001 804-192-R200
89153-830EA 404 USD InStock
89153-830 89153-828
Anti-DNA Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgM

  • For Immunocytochemistry, Immunohistochemistry

Recognizes deoxycytidine of ssDNA of at least 25-30 bases in length in the absence of any reactivity to double-stranded DNA; not species-specific. Specifically detects apoptotic cells; necrotic cells are not recognized.Optimal conditions must be determined individually for each application.

Ordering information: Quantity: 200µl/20 tests; 1ml/100 tests

Type: Primary
Antigen: DNA
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: pan

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty