Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Ceramide Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
804-196-T050
89153-838EA 717 USD InStock
89153-838
Anti-Ceramide Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgM

  • For ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry

Recognizes C16- and C24-ceramide, dihydroceramide, sphingomyelin and phosphatidylcholine in highly artificial lipid overlay test systems. Under more physiological in vitro and in vivo conditions highly specific for ceramide and does not cross-react with sphingomyelin, cholesterol or other phospholipids.

Ordering information: Quantity: 1ml (50 tests). Suggested amount: 20µl/test.

Type: Primary
Antigen: Ceramide
Clonality: Monoclonal
Clone: 0
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty