Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-VASP Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Enzo Life Sciences
804-240F-C100
89153-910EA 1020 USD InStock
89153-910
Anti-VASP Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Host: Mouse, Isotype: IgG1

  • For Immunocytochemistry

Recognizes human, mouse, rat and rabbit Ser239-phosphorylated VASP. Does not cross-react with the non-phosphorylated form of VASP.

Type: Primary
Antigen: VASP
Clonality: Monoclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty