Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CASP8 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
804-242-C100
89153-914EA 536 USD InStock
89153-914
Anti-CASP8 Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG2b

  • For Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human procaspase-8 and active human caspase-8. Detects procaspase-8 (p55/54), the intermediate cleavage products (p43/41) and the p18 active subunit of caspase-8 by Western blot. Does not recognize mouse caspase-8.

Type: Primary
Antigen: CASP8
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty