Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD40L Hamster Monoclonal Antibody

Supplier: Enzo Life Sciences
804-331-C500 804-331-C100
89154-052EA 381 USD InStock
89154-052 89154-054
Anti-CD40L Hamster Monoclonal Antibody
Antibodies
Host: Hamster, Isotype: IgG1

Recognizes mouse CD40L.

Type: Primary
Antigen: CD40L
Clonality: Monoclonal
Clone: 0
Conjugation:
Epitope:
Host: Hamster
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty