Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DLC1 Rat Monoclonal Antibody

Supplier: Enzo Life Sciences
804-340-C100
89154-068EA 481 USD InStock
89154-068
Anti-DLC1 Rat Monoclonal Antibody
Antibodies
Host: Rat, Isotype: IgM

  • For Western Blot

Recognizes human and mouse DLC1. Does not cross-react with DLC2. For detection of both DLC1 and DLC2 see MAb to DLC1 and DLC2 (11F7) (Prod. No. ALX-804-341).

Type: Primary
Antigen: DLC1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgM
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty