Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TLR3 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin))

Supplier: Enzo Life Sciences
804-362R-C100
89154-116EA 729 USD InStock
89154-116
Anti-TLR3 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin))
Antibodies
Host: Mouse, Isotype: IgG1

  • For Immunocytochemistry

Recognizes human, mouse and dog TLR3.

Type: Primary
Antigen: TLR3
Clonality: Monoclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty