Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AHR Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
804-421-R100
89154-246EA 828.89 USD InStock
89154-246
Anti-AHR Mouse Monoclonal Antibody
Antibodies
The aryl hydrocarbon receptor (AhR), also known as the dioxin receptor, is a ligand-activated helix/loop/helix transcription factor found in a variety of vertebrate species. The known ligands for AhR are foreign planar aromatic compounds, such as polycyclic aromatic compounds and halogenated aromatic compounds such as 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD). Unlike the steroid/thyroid hormone receptors, there is no known physiological ligand for the Ah Receptor.
Studies indicate that in non-ligand activated cells, AhR is found complexed with HSP90 predominantly in the cytoplasm. Upon binding to an agonist, the ligand-activated AhR is believed to transform to a nuclear, DNA binding form. This transformation process appears to involve dissociation of HSP90 from AhR followed by formation of a heterodimer with AhR nuclear translocator protein (Arnt). The AhR-ligand complex appears to initiate gene transcription of cytochrome P450 1A1.

  • For Immunohistochemistry, Western Blot

Recognizes human, mouse and rat AhR. Detects a band of ~95kDa by Western blot.

Type: Primary
Antigen: AHR
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty