Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CASP11 Rat Monoclonal Antibody

Supplier: Enzo Life Sciences
804-494-C100
89154-392EA 529 USD InStock
89154-392
Anti-CASP11 Rat Monoclonal Antibody
Antibodies
Caspase-11 not only activates caspase-1, that is required for the maturation of proinflammatory cytokines such as interleukin-1 (IL-1) and interleukin-18 (IL-18), but also activates caspase-3, leading to cellular apoptosis under pathological conditions. In most cells, caspase-11 is only expressed upon induction with pro-inflammatory stimuli. Caspase-11 is an essential mediator of endotoxic shock and caspase-11-deficient mice are resistant to endotoxic shock. LPS-induced caspase-11 regulates lymphocyte apoptosis by activating both caspase-3 and caspase-7. Human caspase-4, thought to be the homolog of mouse caspase-11, may be an effective therapeutic target for treatment of septic shock.

  • For Western Blot

Recognizes mouse and rat caspase-11 (epitope in the p20 fragment). Detects two bands of ~38kDa and ~43kDa by Western blot.

Type: Primary
Antigen: CASP11
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty