Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD14 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
804-496-C100
89154-396EA 513 USD InStock
89154-396
Anti-CD14 Mouse Monoclonal Antibody
Antibodies
The CD14 glycoprotein, gp 55, is present on most monocytic and macrophage like cell types: monocytes, macrophages, Kupffer cells, pleural phagocytic cells and dendritic reticular cells. CD14 is also observed on granulocytes and activated or transformed B-cells. Furthermore CD14 is present in a soluble form in human serum, urine and other body fluids. The CD14 molecule has been reported to be a receptor for endotoxin.

  • For ELISA, Flow Cytometry, Western Blot

Recognizes human (epitope aa 9-13 & aa 39-44), pig, dog, horse and bovine CD14.

Reconstitute with 100µl distilled water.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty