Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL13R Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
804-506-C100
89154-416EA 481 USD InStock
89154-416
Anti-IL13R Mouse Monoclonal Antibody
Antibodies
IL-13 is an immunoregulatory cytokine secreted predominantly by activated TH2 cells, as well as mast cells and basophils, and is a key mediator in the pathogenesis of allergic inflammation. The human IL-13 receptor (IL-13R) is a heterodimer composed of the interleukin-4 receptor a chain (IL-4Rα) and an IL-13 binding protein, IL-13Rα1.A second IL-13 binding protein, IL-13Rα2, has also been identified, which is believed to act as a "decoy" receptor. IL-13 receptors are expressed on human B cells, basophils, eosinophils, mast cells, endothelial cells, fibroblasts, monocytes, macrophages, respiratory epithelial cells, and smooth muscle cells. However, functional IL-13 receptors have not been demonstrated on human or mouse T cells.

Recognizes human IL-13R.

Type: Primary
Antigen: IL13R
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty