Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BMF Rat Monoclonal Antibody

Supplier: Enzo Life Sciences
804-508-C100
89154-420EA 529 USD InStock
89154-420
Anti-BMF Rat Monoclonal Antibody
Antibodies
Host: Rat, Isotype: IgG2a

  • For ELISA, Flow Cytometry, Immunoprecipitation, Western Blot

Recognizes mouse and rat Bmf. Detects bands of ~25kDa and ~30kDa by Western blot, corresponding to endogenous Bmf and BmfL. Does not cross-react with human Bmf.

Type: Primary
Antigen: BMF
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty