Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSP25 Mouse Monoclonal Antibody [clone: 5B9]

Supplier: Enzo Life Sciences
804-588-C100
89154-590EA 525 USD InStock
89154-590
Anti-HSP25 Mouse Monoclonal Antibody [clone: 5B9]
Antibodies
Host: Mouse, Isotype: IgG2a

  • For Immunohistochemistry, Western Blot

Recognizes human, mouse, rat and dog phosphorylated HSP27.

Reconstitute with 1ml water (15 minutes at room temperature).

Type: Primary
Antigen: HSP25
Clonality: Monoclonal
Clone: 5B9
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty