Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TRIP-BR1 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
804-645-C100 804-645-C050
89154-696EA 547 USD InStock
89154-696 89154-694
Anti-TRIP-BR1 Mouse Monoclonal Antibody
Antibodies
TRIP-Br1 (SEI-1) functions as physiological integrator of transcriptional regulatory signals at E2F-responsive promoters to ensure proper execution of E2F-dependent cell cycle progression. TRIP-Br1 is defined as an oncogene.

  • For Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse, pig and monkey TRIP-Br1.

Type: Primary
Antigen: TRIP-BR1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty