Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TACI Rat Monoclonal Antibody

Supplier: Enzo Life Sciences
804-800-C100
89154-994EA 586 USD InStock
89154-994
Anti-TACI Rat Monoclonal Antibody
Antibodies
Host: Rat, Isotype: IgG2a

  • For Immunohistochemistry

Recognizes mouse TACI.

Type: Primary
Antigen: TACI
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty