Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-UNC5B Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
804-846-C100
89155-126EA 513 USD InStock
89155-126
Anti-UNC5B Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG2b

Recognizes human and mouse UNC5B.

Type: Primary
Antigen: UNC5B
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty