Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CEACAM19 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
805-089-C100
89155-390EA 731 USD InStock
89155-390
Anti-CEACAM19 Mouse Monoclonal Antibody
Antibodies
CEA-related cell adhesion molecule 19 (CEACAM19) belongs to the carcinoembryonic antigen (CEA) gene family (1). It encodes a putative glycoprotein which is membrane-bound via a transmembrane domain. Like all members of the CEACAM family, the CEACAM19 protein is a single-pass type I membrane protein and consists of a single extracellular N domain. Two isoforms of CEA-CAM19 exist as a result of alternative splicing events. CEACAM19 expression is ubiquitous with highest expression in prostate, uterus, fetal brain, mammary gland, adrenal gland, skeletal muscle, small intestine, and kidney. Additionally, CEACAM19 is highly expressed in breast and prostate cancer cell lines.

  • For ELISA, Flow Cytometry

Recognizes human CEACAM19. Does not cross-react with human CEACAM1, 3, 4, 5, 6, 7, 8, 20, or 21.

Type: Primary
Antigen: CEACAM19
Clonality: Monoclonal
Clone: 0
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty