Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MT Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
810-014-R100
89155-512EA 810 USD InStock
89155-512
Anti-MT Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit, Isotype: IgG

  • For ELISA, Flow Cytometry, Western Blot

Recognizes human metallothionein.

Type: Primary
Antigen: MT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty