Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DNA (Deoxyribonucleic acid) (from calf thymus) ≥99%, purified

Supplier: Enzo Life Sciences

840-040-C010
89155-846EA 106 USD InStock
89155-846
DNA (Deoxyribonucleic acid) (from calf thymus) ≥99%, purified
DNA (Deoxyribonucleic acid)
Purified from calf thymus and activated by digestion with DNAse I.

For use with PARP-1 or PARP-2 in vitro activity tests

Concentration: 0,2 mg/ml in TRIS-EDTA.


Storage Temperature: Refrigerator
MDL Number: MFCD00130922
CAS Number: 9007-49-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty