Would you like to log in again?
Annexin V-FITC Apoptosis Detection Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
850-020-KI02 850-020-KI01
89155-878EA 550 USD InStock
89155-878 89155-876
Annexin V-FITC Apoptosis Detection Kit, Enzo Life Sciences
Assays Protein Assays
For the detection of apoptotic cells by flow cytometry.
FITC-labelled recombinant human annexin V shows bright green fluorescence (Ex(max): 488 nm, Em(max): 530 nm).
Delivery information: Kit contains Annexin V-FITC, binding buffer and propidium iodide.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty