Would you like to log in again?
Cu/Zn-Superoxide Dismutase ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
850-033-KI01
89155-894EA 1040 USD InStock
89155-894
Cu/Zn-Superoxide Dismutase ELISA Kit, Enzo Life Sciences
Assays ELISAs
For the quantitative determination of human Cu/Zn SOD in cell culture supernatants, serum and other body fluids.
  • Assay Time: <2 hours
Late stages of apoptosis are characterized by the loss of membrane integrity and a release of the intracellular Cu/Zn SOD.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty