Would you like to log in again?
Cell Counting Kit-8, Enzo Life Sciences
Supplier: Enzo Life Sciences
850-039-0100 850-039-KI02 850-039-KI01
89155-900EA 720 USD InStock
89155-900 76199-140 89155-898
Cell Counting Kit-8, Enzo Life Sciences
Assays Cellular Assays
Colorimetric kit for simple and accurate cell proliferation and cytotoxicity assays.
Advantages: One-step, ready-to-use solution, no radioisotopes, no solubilization step, high sensitivity, correlates with the [3H]-thymidine incorporation assay. Can be added directly to the cell media for fast, high-throughput screening without a solubilization process obtaining highly reproducible and accurate results.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty