Would you like to log in again?
Caspase-3 Fluorometric Assay Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
850-216-KI01
89155-960EA 389.33 USD InStock
89155-960
Caspase-3 Fluorometric Assay Kit, Enzo Life Sciences
Assays Protein Assays
Based on the detection of AFC (7-amino-4-trifluoromethyl coumarin) after cleavage from the labelled substrate DEVD-AFC.
Delivery information: Cell Lysis Buffer (25 mL), 2X Reaction Buffer (2 mL), 1 mM DEVD-AFC (125 µL), 1 M DTT (100 µL).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty