Would you like to log in again?
Caspase-8 Colorimetric Assay Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
850-221-KI01
89155-970EA 272.43 USD InStock
89155-970
Caspase-8 Colorimetric Assay Kit, Enzo Life Sciences
Assays Protein Assays
Based on the spectrophotometric detection of the chromophore pNA (p-nitroaniline) after cleavage from the labelled substrate IETD-pNA.
Ordering information: Contents: 25ml Cell Lysis Buffer, 2ml 2X Reaction Buffer, 125µl IETD-pNA Substrate (4mM), 100µl DTT (1M), 25ml Dilution Buffer.
Delivery information: Kit includes Cell lysis buffer (25 mL), 2X Reaction buffer(2 mL), 4 mM IETD-pNA Substrate (125 µL), 1 M DTT (100 µL), Dilution bBuffer (25 mL).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty