Would you like to log in again?
Apoptosis inducer set, Enzo Life Sciences
Supplier: Enzo Life Sciences
850-235-KI01
89155-990EA 578 USD InStock
89155-990
Apoptosis inducer set, Enzo Life Sciences
Assays Cellular Assays
The set contains 5 ready-to-use reagents that induce apoptosis through different mechanisms. Actinomycin D, an antineoplastic antibiotic, inhibits RNA synthesis. Camptothecin, an inhibitor of nuclear topoisomerase, induces apoptosis in many types of cells. Cycloheximide, an active antibiotic against many yeast and fungi, inhibits protein synthesis. Dexamethasone, an active and highly stable glucocorticoid, probably induces apoptosis by binding and activating the intracellular glucocorticoid receptor. Etoposide, a derivative of podophyllotoxin, inhibits topoisomerase activity.
Ordering information: Contents: Contains 5 ready-to-use reagents:
- Actinomycin D, 50µl (10mM), violet cap
- Camptothecin, 1ml (2mM), red cap
- Cycloheximide, 1ml (100mM), green cap
- Dexamethasone, 1ml (10mM), blue cap
- Etoposide, 100µl (100mM), yellow cap
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty