Would you like to log in again?
Caspase inhibitor set lV, Enzo Life Sciences
Supplier: Enzo Life Sciences
850-239-KI01
89155-998EA 1181.69 USD InStock
89155-998
Caspase inhibitor set lV, Enzo Life Sciences
Cell Culture Supplements and Additives
Potent, cell permeable synthetic peptides that irreversibly inhibit activities of each of the caspase-family proteases. YVAD, VDVAD, DEVD, LEVD, WEHD, VEID, IETD, LEHD, and AEVD are the recognition sequences for caspase-1, -2, -3, -4, -5, -6, -8, -9, and -10. VAD is a general caspase inhibitor that inhibits all known caspases. Z-FA-FMK is a negative control for FMK inhibitors.
Ordering information: Contents: Contains 25µl (2mM) each of the following inhibitors:
- Caspase-1 Inhibitor, Z-YVAD-FMK, yellow cap
- Caspase-2 Inhibitor, Z-VDVAD-FMK, orange cap
- Caspase-3 Inhibitor, Z-DEVD-FMK, red cap
- Caspase-4 Inhibitor, Z-LEVD-FMK, orange cap
- Caspase-5 Inhibitor, Z-WEHD-FMK, clear cap
- Caspase-6 Inhibitor, Z-VEID-FMK, violet cap
- Caspase-8 Inhibitor, Z-IETD-FMK, green cap
- Caspase-9 Inhibitor, Z-LEHD-FMK, blue cap
- Caspase-10 Inhibitor, Z-AEVD-FMK, amber cap
- Caspase-13 Inhibitor, Z-LEED-FMK, yellow cap
- Caspase-Family Inhibitor, Z-VAD-FMK, amber cap
- FMK Negative Control, Z-FA-FMK, clear cap
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty