Would you like to log in again?
Recombinant Caspase (active) set lll
Supplier: Enzo Life Sciences
850-243-KI01
89156-004EA 1508.72 USD InStock
89156-004
Recombinant Caspase (active) set lll
Enzymes
We recommend using 1 U per assay for analyzing caspase activity.
Active recombinant caspases are useful in screening caspase inhibitors, studying enzyme kinetics and regulation, determining substrate specificity, and as a positive control in caspase activity assays. 
Ordering information: Contents: Contains 25 units each of the following caspases:
- Caspase-1 (active) (human) (recombinant), yellow cap
- Caspase-2 (active) (human) (recombinant), orange cap
- Caspase-3 (active) (human) (recombinant), red cap
- Caspase-6 (active) (human) (recombinant), violet cap
- Caspase-7 (active) (human) (recombinant), red cap
- Caspase-8 (active) (human) (recombinant), green cap
- Caspase-9 (active) (human) (recombinant), blue cap
- Caspase-10 (active) (human) (recombinant), amber cap
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty