Would you like to log in again?
Cell Counting Kit-F, Enzo Life Sciences
Supplier: Enzo Life Sciences
850-245-KI02 850-245-KI01
89156-010EA 242 USD InStock
89156-010 89156-012
Cell Counting Kit-F, Enzo Life Sciences
Assays Cellular Assays
Fluorometric kit for simple and accurate cell proliferation and cytotoxicity assays
Advantages includes no radioisotopes, more sensitive than the colorimetric assay, short staining reaction time, no solubilization step, correlates with the [3H]-thymidine incorporation assay.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty