Would you like to log in again?
ApoSENSOR™ ADP/ATP Ratio Assay Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
850-248-KI01
89156-018EA 941 USD InStock
89156-018
ApoSENSOR™ ADP/ATP Ratio Assay Kit, Enzo Life Sciences
Assays Cellular Assays
The assay is designed to detect ADP/ATP ratio. The changes in ADP/ATP ratios have been used to conveniently differentiate apoptosis from necrosis.
The assay utilizes bioluminescent detection of ADP and ATP levels for rapid screening of apoptosis (TUNEL-based techniques and caspase assays), necrosis, growth arrest and cell proliferation simultaneously in mammalian cells
This highly sensitive assay (detects 100 mammalian cells/well) can be fully automated for high throughput measurements.
The assay utilizes the enzyme luciferase to catalyze the formation of light fom ATP and luciferin. The light can be measured using a luminometer or beta counter. ADP levels are measured by its conversion to ATP that is subsequently detected using the same reaction.
Delivery information: Kit includes Nucleotide Releasing Buffer (50 mL), 1 vial ATP Monitoring Enzyme (green cap), (1 vial) ADP Monitoring Enzyme (blue cap), 450 µL Enzyme Reconstitution Buffer (red cap).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty