Would you like to log in again?
Mitochondria/Cytosol Fractionation Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
850-276-KI01
89156-082EA 698 USD InStock
89156-082
Mitochondria/Cytosol Fractionation Kit, Enzo Life Sciences
Assays Cellular Assays
For the effective isolation of a highly enriched mitochondrial fraction from the cytosolic fraction of various mammalian cells including both apoptotic an non-apoptotic cells
The core kit contains unique formulations of buffers and reagents that allow isolation of mitochondria and cytosol in only two easy steps. No ultracentrifugations are required. No toxic chemicals are involved.
The enriched mitochondrial and cytosolic fractions can be used to study apoptosis and signal trasduction pathways by detecting translocations of factors of interest between the two fractions by ELISA, Western blot or other assays.
Delivery information: Kit includes Mitochondria Extraction Buffer (10 mL), 5X Cytosol Extraction Buffer (20 mL), 1 M DTT (110 µL), Protease Inhibitor Cocktail (1 vial).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty