Would you like to log in again?
Cytochrome C Releasing Apoptosis Assay Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
850-277-KI01
89156-084EA 924 USD InStock
89156-084
Cytochrome C Releasing Apoptosis Assay Kit, Enzo Life Sciences
Assays Cellular Assays
For the detection of Cytochrome c translocation from mitochondria into cytosol during apoptosis.
The kit provides an easy-to-use procedure and reagents for separating a mitochondria-enriched fraction from cytosol. Cytochrome c released from mitochondria into cytosol is determined by Western blot using the cytochrome c antibody provided in the kit. The procedure is simple, straightforward and requires no ultracentrifugation.
Delivery information: Kit includes Mitochondria Extraction Buffer (10 mL), 5X Cytosol Extraction Buffer (20 mL), 1 M DTT (110 µL), 500X Protease Inhibitor Cocktail (1 vial), 0.2 mg/mL Cytochrome c Antibody (0.5 mL).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty