Would you like to log in again?
OXI-TEK TBARS Assay Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
850-287-KI01
89156-088EA 924 USD InStock
89156-088
OXI-TEK TBARS Assay Kit, Enzo Life Sciences
Assays Cellular Assays
The sensitivity of measuring thiobarbituric acid reactive substances (TBARS) has made this assay the method of choice for screening and monitoring lipid peroxidation, which is a major indication of oxidative stress.
Malondialdehyde (MDA) forms a 1:2 adduct with thiobarbituric acid; the adduct can be measured by fluorometry. Biological specimens contain a mixture of TBARS, including lipid hydroperoxides and aldehydes. In practice, TBARS are expressed in equivalent numbers to MDA.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty